Learn More
Biomatik Corporation Human Leukocyte Associated Immunoglobulin Like Receptor 1 (LAIR1) ELISA Kit
Supplier:  Biomatik Corporation EKU05619
Encompass_Preferred
Human Leukocyte Associated Immunoglobulin Like Receptor 1 (LAIR1) ELISA Kit for detecting LAIR1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6GTX8. Gene ID: 3903. Target Synonyms: CD305; hLAIR1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2615