Learn More
Biomatik Corporation Human Low Density Lipoprotein Receptor Related Protein 6 (LRP6) ELISA Kit
Supplier:  Biomatik Corporation EKU05685
Encompass_Preferred
Sample Type: Tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.056ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: O75581. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-2624