Learn More
Biomatik Corporation Simian Luteinizing Hormone (LH) ELISA Kit
Supplier:  Biomatik Corporation EKU05696
Encompass_Preferred
Simian Luteinizing Hormone (LH) ELISA Kit for detecting LH in serum, plasma, other biological fluids. Species Reactivity: Simian. Detection Range: 370.4-30,000pg/ml. Sensitivity: 137.2pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q6EV78. Target Synonyms: ICSH; Lutropin; Interstitial Cell Stimulating Hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2627