Learn More
Biomatik Corporation Human Lysophosphatidic Acid Receptor 3 (LPAR3) ELISA Kit
Supplier:  Biomatik Corporation EKU05712
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.113ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: GPCR; EDG7; HOFNH30; LP-A3; LPA3; LPAR3; Endothelial Differentiation,Lysophosphatidic Acid G-Protein-Coupled Receptor 7; Lysophosphatidic acid receptor Edg-7. Restriction: For Research Use Only.
Catalog No. 50-229-2631