Learn More
Biomatik Corporation Human Lysyl tRNA Synthetase (KARS) ELISA Kit
Supplier:  Biomatik Corporation EKU05737
Encompass_Preferred
Sample Type: Serum, plasma, tissue homogenatesand other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.127ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q15046. Target Synonyms: KARS2; LysRS; Lysine tRNA Ligase. Restriction: For Research Use Only.
Catalog No. 50-229-2633