Learn More
Biomatik Corporation Human Macrophage Inflammatory Protein 5 (MIP5) ELISA Kit
Supplier:  Biomatik Corporation EKU05760
Encompass_Preferred
Human Macrophage Inflammatory Protein 5 (MIP5) ELISA Kit for detecting MIP5 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q16663. Gene ID: 6359. Target Synonyms: CCL15; SCYA15; HCC-2; NCC-3; SCYL3; Lkn-1; MIP1-D; MIP1d; MIP-1d; HMRP-2B; Leukotactin-1; Chemokine C-C-Motif Ligand 15; MIP-1 Delta; Small-inducible cytokine A15. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2637