Learn More
Biomatik Corporation Human Malate Dehydrogenase 2 (MDH2) ELISA Kit
Supplier:  Biomatik Corporation EKU05781
Encompass_Preferred
Human Malate Dehydrogenase 2 (MDH2) ELISA Kit for detecting MDH2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.61ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P40926. Target Synonyms: M-MDH; MDH; MOR1; Malate Dehydrogenase 2,NAD (Mitochondrial). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2639