Learn More
Biomatik Corporation Human Acid sphingomyelinase-like phosphodiesterase 3b (SMPDL3B) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC32460
Encompass_Preferred
The Human Acid sphingomyelinase-like phosphodiesterase 3b (SMPDL3B) ELISA Kit allows for quantitative determination of sphingomyelin phosphodiesterase, acid-like 3B in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human. Detection Range: 0.625 ng/mL-40 ng/mL. Sensitivity: 0.156 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q92485. Restriction: For Research Use Only.
Catalog No. 50-149-7646