Learn More
Biomatik Corporation Horse Matrix Metalloproteinase 2 (MMP2) ELISA Kit
Supplier:  Biomatik Corporation EKU05849
Encompass_Preferred
Horse Matrix Metalloproteinase 2 (MMP2) ELISA Kit for detecting MMP2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Horse (Equus caballus; Equine). Detection Range: 1.56-100ng/ml. Sensitivity: 0.65ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: F7AYK2. Target Synonyms: CLG4; CLG4A; MMP-II; MONA; TBE-1; PEX; Gelatinase A; 72kDa Gelatinase; 72kDa Type IV Collagenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2655