Learn More
Biomatik Corporation Horse Matrix Metalloproteinase 3 (MMP3) ELISA Kit
Supplier:  Biomatik Corporation EKU05862
Encompass_Preferred
Horse Matrix Metalloproteinase 3 (MMP3) ELISA Kit for detecting MMP3 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Horse (Equus caballus; Equine). Detection Range: 31.2-2,000pg/ml. Sensitivity: 11.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q28397. Gene ID: 100034195. Target Synonyms: SL1; STMY; STMY1; STR1; Progelatinase; Stromelysin 1; Transin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2660