Learn More
Biomatik Corporation Rabbit Matrix Metalloproteinase 3 (MMP3) ELISA Kit
Supplier:  Biomatik Corporation EKU05867
Encompass_Preferred
Rabbit Matrix Metalloproteinase 3 (MMP3) ELISA Kit for detecting MMP3 in serum, plasma, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.119ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P28863. Gene ID: 100009111. Target Synonyms: SL1; STMY; STMY1; STR1; Progelatinase; Stromelysin 1; Transin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2662