Learn More
Biomatik Corporation Mouse Microtubule Associated Protein 2 (MAP2) ELISA Kit
Supplier:  Biomatik Corporation EKU05951
Encompass_Preferred
Mouse Microtubule Associated Protein 2 (MAP2) ELISA Kit for detecting MAP2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P20357. Gene ID: 17756. Target Synonyms: MAP2A; MAP2B; MAP2C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2675