Learn More
Biomatik Corporation Human Myosin Light Chain 9, Regulatory (MYL9) ELISA Kit
Supplier:  Biomatik Corporation EKU06108
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenatesand other biological fluids. Species Reactivity: Human. Detection Range: 78-5,000pg/mL. Sensitivity: 33pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P24844. Target Synonyms: MLC2; LC20; MYRL2; MRLC1; 20 kDa myosin light chain; Myosin Regulatory Light Chain 2,Smooth Muscle Isoform; Myosin Regulatory Light Chain 1. Restriction: For Research Use Only.
Catalog No. 50-229-2698