Learn More
Biomatik Corporation Rat Myristoylated Alanine Rich Protein Kinase C Substrate (MARCKS) ELISA Kit
Supplier:  Biomatik Corporation EKU06124
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids.. Species Reactivity: Rat. Detection Range: 1.56-100ng/mL. Sensitivity: 0.54ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: 80K-L; MACS; MRACKS; PKCSL; PRKCSL; Protein kinase C substrate, 80 kDa protein, light chain. Restriction: For Research Use Only.
Catalog No. 50-229-2700