Learn More
Biomatik Corporation Human NADH Dehydrogenase Ubiquinone Fe-S Protein 1 (NDUFS1) ELISA Kit
Supplier:  Biomatik Corporation EKU06146
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.114ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P28331. Target Synonyms: CI-75Kd; NADH-Coenzyme Q Reductase; NADH Dehydrogenase Iron-Sulfur Protein 1; NADH-ubiquinone oxidoreductase 75 kDa subunit, mitochondrial. Restriction: For Research Use Only.
Catalog No. 50-229-2705