Learn More
Biomatik Corporation Bovine Nerve Growth Factor (NGF) ELISA Kit
Supplier:  Biomatik Corporation EKU06171
Encompass_Preferred
Bovine Nerve Growth Factor (NGF) ELISA Kit for detecting NGF in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P13600. Gene ID: 281350. Target Synonyms: NGFB; Beta-NGF; HSAN5; NGF-B; Beta-Nerve Growth Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2709