Learn More
Biomatik Corporation Rat Nerve Growth Factor IB (NGFIB) ELISA Kit
Supplier:  Biomatik Corporation EKU06179
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.156-10ng/mL. Sensitivity: 0.061ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P22829. Target Synonyms: GFRP1; HMR; N10; NAK-1; NGFIB; NP10; NUR77; TR3; DHR38; TIS1; Orphan nuclear receptor HMR; Testicular receptor 3; Nuclear receptor subfamily 4 group A member 1. Restriction: For Research Use Only.
Catalog No. 50-229-2712