Learn More
Biomatik Corporation Rat Neurocan (NCAN) ELISA Kit
Supplier:  Biomatik Corporation EKU06207
Encompass_Preferred
Rat Neurocan (NCAN) ELISA Kit for detecting NCAN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P55067. Target Synonyms: CSPG3; NEUR; Neurocan Core Protein; Chondroitin Sulfate Proteoglycan 3; Neurocan Proteoglycan. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2716