Learn More
Biomatik Corporation Mouse Neuronal Pentraxin I (NPTX1) ELISA Kit
Supplier:  Biomatik Corporation EKU06235
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.057ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q62443. Target Synonyms: NARP1; NP-I; NP1. Restriction: For Research Use Only.
Catalog No. 50-229-2718