Learn More
Biomatik Corporation Rat Neuropeptide S (NPS) ELISA Kit
Supplier:  Biomatik Corporation EKU06244
Encompass_Preferred
Rat Neuropeptide S (NPS) ELISA Kit for detecting NPS in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 6.4-4,000pg/ml. Sensitivity: 2.5pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P0C0P7. Gene ID: 100360071. Target Synonyms: NP-S. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2720