Learn More
Biomatik Corporation Mouse Neurotrophin 4 (NT4) ELISA Kit
Supplier:  Biomatik Corporation EKU06274
Encompass_Preferred
Mouse Neurotrophin 4 (NT4) ELISA Kit for detecting NT4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q80VU4. Gene ID: 78405. Target Synonyms: NT5; NTF4; NT-4/5; Neurotrophic Factor 4; Neurotrophin-5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2724