Learn More
Biomatik Corporation Human activated protein C, APC ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC32473
Encompass_Preferred
The Human activated protein C, APC ELISA Kit allows for quantitative determination of activated protein C, APC in serum, plasma, tissue homogenates. Species Reactivity: Human. Detection Range: 1.56 pg/mL-100 pg/mL. Sensitivity: 0.39 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. Restriction: For Research Use Only.
Catalog No. 50-149-7649