Learn More
Biomatik Corporation Human Oligodendrocyte Lineage Transcription Factor 2 (OLIG2) ELISA Kit
Supplier:  Biomatik Corporation EKU06388
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 15.6-1,000pg/mL. Sensitivity: 5.9pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: BHLHB1; OLIGO2; PRKCBP2; RACK17; Basic Domain,Helix-Loop-Helix Protein,Class B 1; Class B basic helix-loop-helix protein 1; Class E basic helix-loop-helix protein 19. Restriction: For Research Use Only.
Catalog No. 50-229-2739