Learn More
Biomatik Corporation Rabbit Osteopontin (OPN) ELISA Kit
Supplier:  Biomatik Corporation EKU06434
Encompass_Preferred
Sample Type: Serum, plasma, tissue homogenatesand other biological fluids. Species Reactivity: Rabbit. Detection Range: 0.156-10ng/mL. Sensitivity: 0.061ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P31097. Target Synonyms: SPP1; BNSP; BSPI; ETA1; Secreted Phosphoprotein 1; Bone Sialoprotein I; Early T-Lymphocyte Activation 1; Nephropontin; Urinary stone protein; Uropontin. Restriction: For Research Use Only.
Catalog No. 50-229-2745