Learn More
Biomatik Corporation Human p21 Protein Activated Kinase 4 (PAK4) ELISA Kit
Supplier:  Biomatik Corporation EKU06457
Encompass_Preferred
Human p21 Protein Activated Kinase 4 (PAK4) ELISA Kit for detecting PAK4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O96013. Gene ID: 10298. Target Synonyms: Serine/threonine-protein kinase PAK 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2748