Learn More
Biomatik Corporation Human Palladin (PALLD) ELISA Kit
Supplier:  Biomatik Corporation EKU06464
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.054ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q8WX93. Target Synonyms: Cytoskeletal Associated Protein; Sarcoma antigen NY-SAR-77. Restriction: For Research Use Only.
Catalog No. 50-229-2751