Learn More
Biomatik Corporation Human Parkinson Disease Protein 2 (PARK2) ELISA Kit
Supplier:  Biomatik Corporation EKU06492
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.055ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: O60260. Target Synonyms: AR-JP; LPRS2; PDJ; PRKN; Parkin; Autosomal Recessive,Juvenile; E3 ubiquitin-protein ligase parkin. Restriction: For Research Use Only.
Catalog No. 50-229-2754