Learn More
Biomatik Corporation Rat Plasminogen (Plg) ELISA Kit
Supplier:  Biomatik Corporation EKU06657
Encompass_Preferred
Rat Plasminogen (Plg) ELISA Kit for detecting Plg in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000ng/ml. Sensitivity: 7.2ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q01177. Gene ID: 85253. Target Synonyms: PL; Plasmin; Activation peptide; Angiostatin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2775