Learn More
Biomatik Corporation Human Platelet Derived Growth Factor C (PDGFC) ELISA Kit
Supplier:  Biomatik Corporation EKU06686
Encompass_Preferred
Human Platelet Derived Growth Factor C (PDGFC) ELISA Kit for detecting PDGFC in plasma. Species Reactivity: Human (Homo sapiens). Detection Range: 125.00-8000pg/ml. Sensitivity: 43pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9NRA1. Gene ID: 56034. Target Synonyms: PDGF-C; SCDGF; VEGF-E; Fallotein; Spinal cord-derived growth factor; Platelet-derived growth factor C, latent form; Platelet-derived growth factor C, receptor-binding form. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2778