Learn More
Biomatik Corporation Rat Presenilin 2 (PS2) ELISA Kit
Supplier:  Biomatik Corporation EKU06764
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.156-10ng/mL. Sensitivity: 0.055ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: PSEN2; AD3L; AD4; PS2; STM2; AD3LP; AD5; Alzheimer Disease 4. Restriction: For Research Use Only.
Catalog No. 50-229-2789