Learn More
Biomatik Corporation Mouse Prokineticin 2 (PK2) ELISA Kit
Supplier:  Biomatik Corporation EKU06815
Encompass_Preferred
Mouse Prokineticin 2 (PK2) ELISA Kit for detecting PK2 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 49.4-4,000pg/ml. Sensitivity: 19.1pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q9QXU7. Gene ID: 50501. Target Synonyms: PK-2; Bv8; MIT1; KAL4; Protein Bv8 Homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2793