Learn More
Biomatik Corporation Human Protection Of Telomeres 1 Homolog (POT1) ELISA Kit
Supplier:  Biomatik Corporation EKU06891
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.114ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9NUX5. Target Synonyms: hPot1; POT1-like telomere end-binding protein. Restriction: For Research Use Only.
Catalog No. 50-229-2805