Learn More
Biomatik Corporation Human RAD54 Like Protein 2 (RAD54L2) ELISA Kit
Supplier:  Biomatik Corporation EKU07008
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.057ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9Y4B4. Target Synonyms: SRISNF2L; Helicase ARIP4; Androgen receptor-interacting protein 4. Restriction: For Research Use Only.
Catalog No. 50-229-2826