Learn More
Biomatik Corporation Mouse Regenerating Islet Derived Protein 4 (REG4) ELISA Kit
Supplier:  Biomatik Corporation EKU07050
Encompass_Preferred
Mouse Regenerating Islet Derived Protein 4 (REG4) ELISA Kit for detecting REG4 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 46.88-3,000pg/ml. Sensitivity: 16.29pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9D8G5. Gene ID: 67709. Target Synonyms: GISP; REG-IV; RELP; Regenerating Gene Type IV; Gastrointestinal Secretory Protein; REG-like protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2836