Learn More
Biomatik Corporation Mouse Regulator Of G Protein Signaling 5 (RGS5) ELISA Kit
Supplier:  Biomatik Corporation EKU07057
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids.. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.067ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: O08850. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-2840