Learn More
Biomatik Corporation Human Renal Tumor Antigen (RAGE) ELISA Kit
Supplier:  Biomatik Corporation EKU07069
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.057ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9UQ07. Target Synonyms: MOK; RAGE1; MAPK/MAK/MRK Overlapping Kinase. Restriction: For Research Use Only.
Catalog No. 50-229-2842