Learn More
Biomatik Corporation Bovine Retinol Binding Protein 4 (RBP4) ELISA Kit
Supplier:  Biomatik Corporation EKU07103
Encompass_Preferred
Bovine Retinol Binding Protein 4 (RBP4) ELISA Kit for detecting RBP4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.141ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P18902. Target Synonyms: PRBP; RBP; Plasma retinol-binding protein; Retinol Binding Protein 4, Plasma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2846