Learn More
Biomatik Corporation Rat S100 Calcium Binding Protein A6 (S100A6) ELISA Kit
Supplier:  Biomatik Corporation EKU07174
Encompass_Preferred
Rat S100 Calcium Binding Protein A6 (S100A6) ELISA Kit for detecting S100A6 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P05964. Gene ID: 85247. Target Synonyms: S100-A6; 2A9; 5B10; CABP; CACY; PRA; MLN 4; Calcyclin; Growth factor-inducible protein 2A9; Prolactin receptor-associated protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2856