Learn More
Biomatik Corporation Human Secretogranin III (SCG3) ELISA Kit
Supplier:  Biomatik Corporation EKU07231
Encompass_Preferred
Sample Type: Serum, plasma, tissue homogenatesand other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.062ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q8WXD2. Target Synonyms: SGIII. Restriction: For Research Use Only.
Catalog No. 50-229-2862