Learn More
Biomatik Corporation Human Semaphorin 4B (SEMA4B) ELISA Kit
Supplier:  Biomatik Corporation EKU07266
Encompass_Preferred
Human Semaphorin 4B (SEMA4B) ELISA Kit for detecting SEMA4B in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9NPR2. Gene ID: 10509. Target Synonyms: SEMAC; SEMC; Sema Domain, Transmembrane Domain And Cytoplasmic Domain Protein 4B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2866