Learn More
Biomatik Corporation Human Serine/Threonine Kinase 39 (STK39) ELISA Kit
Supplier:  Biomatik Corporation EKU07292
Encompass_Preferred
Sample Type: Tissue homogenates and other biological fluids.. Species Reactivity: Human. Detection Range: 0.781-50ng/mL. Sensitivity: 0.24ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9UEW8. Target Synonyms: DCHT; SPAK; STE20/SPS1-related proline-alanine-rich protein kinase. Restriction: For Research Use Only.
Catalog No. 50-229-2870