Learn More
Biomatik Corporation Human SHC-Transforming Protein 1 (SHC1) ELISA Kit
Supplier: Biomatik Corporation EKU07321
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.117ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P29353. Target Synonyms: SHC; SHCA; p52SHC; p66; p66SHC; SHC-transforming protein 3; SHC-transforming protein A; Src homology 2 domain-containing-transforming protein C1. Restriction: For Research Use Only.