Learn More
Biomatik Corporation Rat Signal Transducer And Activator Of Transcription 6 (STAT6) ELISA Kit
Supplier:  Biomatik Corporation EKU07345
Encompass_Preferred
Rat Signal Transducer And Activator Of Transcription 6 (STAT6) ELISA Kit for detecting STAT6 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q1KQ07. Gene ID: 362896. Target Synonyms: IL-4-STAT; STAT6B; STAT6C; Signal Transducer And Activator Of Transcription 6,Interleukin-4 Induced. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2880