Learn More
Biomatik Corporation Human Solute Carrier Family 3, Member 2 (SLC3A2) ELISA Kit
Supplier:  Biomatik Corporation EKU07396
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.057ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P08195. Target Synonyms: SLC3A2; CD98HC; 4F2; 4F2HC; 4T2HC; MDU1; NACAE; Activators Of Dibasic And Neutral Amino Acid Transport; Antigen Identified By Monoclonal Antibodies 4F2; CD98 Heavy Chain. Restriction: For Research Use Only.
Catalog No. 50-229-2888