Learn More
Biomatik Corporation Mouse Spermine Oxidase (SMOX) ELISA Kit
Supplier:  Biomatik Corporation EKU07428
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.312-20ng/mL. Sensitivity: 0.115ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: SMO; PAO; PAOh1; C20orf16; Polyamine oxidase 1. Restriction: For Research Use Only.
Catalog No. 50-229-2894