Learn More
Biomatik Corporation Rat Superoxide Dismutase 3, Extracellular (SOD3) ELISA Kit
Supplier:  Biomatik Corporation EKU07506
Encompass_Preferred
Rat Superoxide Dismutase 3, Extracellular (SOD3) ELISA Kit for detecting SOD3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.32ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q08420. Target Synonyms: SOD-3; EC-SOD; Extracellular Superoxide Dismutase Cu-Zn. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2906