Learn More
Biomatik Corporation Rat Suppressors Of Cytokine Signaling 3 (SOCS3) ELISA Kit
Supplier:  Biomatik Corporation EKU07518
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.156-10ng/mL. Sensitivity: 0.057ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: O88583. Target Synonyms: ATOD4; CIS3; Cish3; MGC71791; SOCS-3; SSI-3; SSI3; Cytokine-inducible SH2 protein 3; STAT-induced STAT inhibitor 3. Restriction: For Research Use Only.
Catalog No. 50-229-2909