Learn More
Biomatik Corporation Human Synaptotagmin Like Protein 2 (SYTL2) ELISA Kit
Supplier:  Biomatik Corporation EKU07545
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.117ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9HCH5. Target Synonyms: CHR11SYT; SGA72M; SLP2; Exophilin-4; Breast Cancer-Associated Antigen SGA-72M. Restriction: For Research Use Only.
Catalog No. 50-229-2913