Learn More
Biomatik Corporation Human Telomeric Repeat Binding Factor 1 (TERF1) ELISA Kit
Supplier:  Biomatik Corporation EKU07589
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids.. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.055ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P54274. Target Synonyms: TRF; PIN2; TRBF1; TRF1; hTRF1-AS; t-TRF1; NIMA-interacting protein 2; TTAGGG repeat-binding factor 1; Telomeric protein Pin2/TRF1. Restriction: For Research Use Only.
Catalog No. 50-229-2920