Learn More
Biomatik Corporation Human Testican 2 (TIC2) ELISA Kit
Supplier:  Biomatik Corporation EKU07603
Encompass_Preferred
Human Testican 2 (TIC2) ELISA Kit for detecting TIC2 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q92563. Gene ID: 9806. Target Synonyms: SPOCK2; Sparc/Osteonectin,Swcv And Kazal-Like Domains Proteoglycan 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2924